siRNA Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025115.1 | NP_001020286.1 | 6 | 626 | ||
| XM_006250812.1 | XP_006250874.1 | 6 | 775 | |||
| GenBank mRNA | BC091249.1 | - | 6 | 626 | 7368126 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stap1| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025115.1 | NP_001020286.1 | 6 | 626 | ||
| XM_006250812.1 | XP_006250874.1 | 6 | 775 | |||
| GenBank mRNA | BC091249.1 | - | 6 | 626 | 7368126 | |