siRNA Details
| Gene Symbol: Fam63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025118.1 | NP_001020289.1 | 1 | 198 | ||
| XM_006232886.2 | XP_006232948.1 | 4 | 1422 | |||
| XM_008761304.1 | XP_008759526.1 | 4 | 1422 | |||
| GenBank mRNA | BC091386.1 | - | 1 | 198 | 7321358 | |

Fam63a
Genomic Map