siRNA Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025120.1 | NP_001020291.1 | 16 | 2386 | ||
| GenBank mRNA | BC087088.1 | - | 16 | 2386 | 7190300 | |
| FQ226627.1 | - | 6 | 565 | |||
| FQ230872.1 | - | 14 | 1993 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf3c2| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025120.1 | NP_001020291.1 | 16 | 2386 | ||
| GenBank mRNA | BC087088.1 | - | 16 | 2386 | 7190300 | |
| FQ226627.1 | - | 6 | 565 | |||
| FQ230872.1 | - | 14 | 1993 | |||