siRNA Details
Gene Symbol: RGD1561328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025122.2 | NP_001020293.2 | 2 | 1249 | ||
GenBank mRNA | BC091178.1 | - | 2 | 1249 | 7325408 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1561328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025122.2 | NP_001020293.2 | 2 | 1249 | ||
GenBank mRNA | BC091178.1 | - | 2 | 1249 | 7325408 |