siRNA Details
| Gene Symbol: Rfwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025126.1 | NP_001020297.1 | 3 | 401 | ||
| XM_006250113.2 | XP_006250175.1 | 10 | 1600 | |||
| XM_006250114.2 | XP_006250176.1 | 10 | 1588 | |||
| XM_006250115.2 | XP_006250177.1 | 9 | 1540 | |||
| XM_006250116.2 | XP_006250178.1 | 9 | 1528 | |||
| GenBank mRNA | BC091284.1 | - | 3 | 401 | 7304516 | |

Rfwd2
Genomic Map