siRNA Details
| Gene Symbol: Rufy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025127.1 | NP_001020298.1 | 13, 14 | 1783 | ||
| XM_008770047.1 | XP_008768269.1 | 12, 13 | 1584 | |||
| XM_008770048.1 | XP_008768270.1 | 14, 15 | 2700 | |||
| GenBank mRNA | BC089952.1 | - | 13, 14 | 1783 | 7312240 | |
| EF538802.1 | - | 13, 14 | 1388 | |||
| EF577045.1 | - | 12, 13 | 1334 | |||

Rufy3
Genomic Map