siRNA Details
| Gene Symbol: Fam107a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025129.1 | NP_001020300.1 | 4 | 1061 | ||
| GenBank mRNA | BC093601.1 | - | 4 | 1061 | 7316049 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam107a| Gene Symbol: Fam107a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025129.1 | NP_001020300.1 | 4 | 1061 | ||
| GenBank mRNA | BC093601.1 | - | 4 | 1061 | 7316049 | |