siRNA Details
| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006045.2 | NP_006036.1 | 28 | 4993 | ||
| XM_011528481.2 | XP_011526783.1 | 28 | 5021 | |||
| GenBank mRNA | AB014511.1 | - | 23 | 4549 | ||
| AK025559.1 | - | 1 | 821 | |||
| BC036759.1 | - | 1 | 1271 | |||
| BC110591.1 | - | 24 | 4477 | |||
| BC110592.1 | - | 24 | 4475 | |||

ATP9A
Genomic Map