siRNA Details
| Gene Symbol: Snn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009223.3 | NP_033249.1 | 2 | 2456 | ||
| GenBank mRNA | AK012171.1 | - | 1 | 764 | ||
| AK013035.1 | - | 1 | 842 | |||
| AK149685.1 | - | 2 | 2405 | |||
| AK210118.1 | - | 1 | 183 | |||
| BC006961.1 | - | 2 | 2424 | 3707801 | ||
| BC017685.1 | - | 2 | 2373 | 4241981 | ||
| BC038475.1 | - | 1 | 2497 | 3157762 | ||
| BC063027.1 | - | 2 | 2393 | 5343626 | ||

Snn
Genomic Map