siRNA Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 10, 11 | 1010 | ||
| GenBank mRNA | AK138608.1 | - | 10, 11 | 1011 | ||
| BC141362.1 | - | 8, 9 | 927 | 9055974 | ||
| CR756940.1 | - | 4, 5 | 319 | |||

Myh4
Genomic Map