siRNA Details
| Gene Symbol: Nkx2-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010920.2 | NP_035050.2 | 2 | 1067 | ||
| GenBank mRNA | AF030113.1 | - | 2 | 878 | ||
| AK006401.1 | - | 2 | 923 | |||
| AK007304.1 | - | 2 | 850 | |||
| BC100425.1 | - | 2 | 885 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx2-6| Gene Symbol: Nkx2-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010920.2 | NP_035050.2 | 2 | 1067 | ||
| GenBank mRNA | AF030113.1 | - | 2 | 878 | ||
| AK006401.1 | - | 2 | 923 | |||
| AK007304.1 | - | 2 | 850 | |||
| BC100425.1 | - | 2 | 885 | |||