siRNA Details
| Gene Symbol: Nkx2-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010920.2 | NP_035050.2 | 2 | 677 | ||
| GenBank mRNA | AF030113.1 | - | 2 | 490 | ||
| AF045150.1 | - | 2 | 677 | |||
| AK006401.1 | - | 2 | 533 | |||
| AK007304.1 | - | 2 | 460 | |||
| BC100425.1 | - | 2 | 495 | |||
| BC132558.1 | - | 2 | 527 | 40130835 | ||
| BC139035.1 | - | 2 | 527 | 8862057 | ||
| BC145516.1 | - | 2 | 531 | |||

Nkx2-6
Genomic Map