siRNA Details
| Gene Symbol: Tst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012808.1 | NP_036940.1 | 1 | 324 | ||
| GenBank mRNA | BC088449.1 | - | 1 | 324 | 7126617 | |
| X56228.1 | - | 1 | 287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tst| Gene Symbol: Tst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012808.1 | NP_036940.1 | 1 | 324 | ||
| GenBank mRNA | BC088449.1 | - | 1 | 324 | 7126617 | |
| X56228.1 | - | 1 | 287 | |||