siRNA Details
| Gene Symbol: ZNF623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082480.2 | NP_001075949.1 | 2 | 2142 | ||
| NM_001261843.1 | NP_001248772.1 | 2 | 2021 | |||
| NM_014789.3 | NP_055604.3 | 1 | 2002 | |||
| XM_006716708.3 | XP_006716771.1 | 2 | 2074 | |||
| GenBank mRNA | AB014528.1 | - | 1 | 2126 | ||
| BC112899.1 | - | 1 | 1516 | |||
| DA211779.1 | - | 1 | 334 | |||

ZNF623
Genomic Map