siRNA Details
| Gene Symbol: KIAA0907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014949.3 | NP_055764.2 | 5, 6 | 597 | ||
| GenBank mRNA | AB020714.1 | - | 5, 6 | 554 | ||
| AK289902.1 | - | 5, 6 | 540 | |||
| AY112680.1 | - | 5, 6 | 528 | |||
| AY112681.1 | - | 5, 6 | 528 | |||
| AY112682.1 | - | 5, 6 | 528 | |||
| BC027182.1 | - | 5, 6 | 553 | |||
| BC062637.1 | - | 5, 6 | 552 | 5497581 | ||
| KU178555.1 | - | 5, 6 | 528 | |||
| KU178556.1 | - | 5 | 792 | |||

KIAA0907
Genomic Map