siRNA Details
Gene Symbol: RRP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015056.2 | NP_055871.1 | 16 | 4248 | ||
GenBank mRNA | AK124620.1 | - | 5 | 3797 | ||
D80001.1 | - | 16 | 4156 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RRP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015056.2 | NP_055871.1 | 16 | 4248 | ||
GenBank mRNA | AK124620.1 | - | 5 | 3797 | ||
D80001.1 | - | 16 | 4156 |