siRNA Details
| Gene Symbol: VWA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009814.1 | NP_001009814.1 | 3 | 351 | ||
| NM_015058.1 | NP_055873.1 | 3 | 351 | |||
| XM_011535007.2 | XP_011533309.1 | 3 | 377 | |||
| XM_017020469.1 | XP_016875958.1 | 5 | 410 | |||
| XM_017020470.1 | XP_016875959.1 | 3 | 377 | |||
| XM_017020471.1 | XP_016875960.1 | 3 | 377 | |||
| XM_017020474.1 | XP_016875963.1 | 3 | 377 | |||
| GenBank mRNA | AK095155.1 | - | 3 | 225 | ||
| BC053674.1 | - | 3 | 351 | 6171968 | ||
| BC131802.1 | - | 4 | 184 | |||

VWA8
Genomic Map