siRNA Details
| Gene Symbol: DDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015086.1 | NP_055901.2 | 2 | 2766 | ||
| XM_011538055.2 | XP_011536357.1 | 3 | 3371 | |||
| GenBank mRNA | AB018292.1 | - | 2 | 2650 | ||
| BC136398.1 | - | 2 | 2676 | 9020385 | ||
| BC136399.1 | - | 2 | 2676 | 9020386 | ||

DDN
Genomic Map