siRNA Details
| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 8 | 560 | ||
| GenBank mRNA | CN297641.1 | - | 8 | 557 | ||
| D79991.1 | - | 8 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUP188| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 8 | 560 | ||
| GenBank mRNA | CN297641.1 | - | 8 | 557 | ||
| D79991.1 | - | 8 | 522 | |||