siRNA Details
| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 24 | 2520 | ||
| GenBank mRNA | BC005407.1 | - | 5 | 461 | ||
| BC040352.1 | - | 11 | 1127 | |||
| BC111045.1 | - | 2 | 169 | |||
| BX537774.1 | - | 21 | 2164 | |||
| D79991.1 | - | 24 | 2482 | |||

NUP188
Genomic Map