siRNA Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018046.4 | NP_060516.2 | 11 | 2046 | ||
| GenBank mRNA | AK001145.1 | - | 6 | 558 | ||
| AK289394.1 | - | 7 | 898 | |||
| AY500994.1 | - | 11 | 1965 | |||
| BC015478.1 | - | 6 | 797 | |||
| BC029382.1 | - | 4 | 355 | |||
| BX648120.1 | - | 7 | 855 | |||

AGGF1
Genomic Map