siRNA Details
| Gene Symbol: Nrxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205234.1 | NP_001192163.1 | 19 | 4089 | ||
| NM_001205235.1 | NP_001192164.1 | 18 | 4065 | |||
| NM_020253.3 | NP_064649.2 | 19 | 4089 | |||
| GenBank mRNA | AK129239.1 | - | 19 | 4062 | ||
| AK147295.1 | - | 19 | 4022 | |||
| AK147665.1 | - | 19 | 4021 | |||
| BC145496.1 | - | 17 | 3712 | 9054059 | ||
| BC150784.1 | - | 17 | 3712 | 9087695 | ||
| BM944179.1 | - | 1 | 41 | |||
| BM950483.1 | - | 2 | 79 | |||

Nrxn2
Genomic Map