siRNA Details
| Gene Symbol: AMIGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020703.2 | NP_065754.2 | 2 | 1505 | ||
| XM_011541812.2 | XP_011540114.1 | 2 | 1511 | |||
| GenBank mRNA | AB032989.1 | - | 1 | 957 | ||
| AK295678.1 | - | 2 | 1484 | |||
| AY237007.1 | - | 1 | 1123 | |||
| BC040879.1 | - | 2 | 1473 | 5725643 | ||

AMIGO1
Genomic Map