siRNA Details
| Gene Symbol: RNF150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020724.1 | NP_065775.1 | 4 | 1512 | ||
| XM_005263150.4 | XP_005263207.1 | 5 | 1026 | |||
| XM_011532147.2 | XP_011530449.1 | 5 | 489 | |||
| XM_011532148.2 | XP_011530450.1 | 5 | 504 | |||
| XM_017008475.1 | XP_016863964.1 | 4 | 423 | |||
| XM_017008476.1 | XP_016863965.1 | 4 | 396 | |||
| GenBank mRNA | AB033040.1 | - | 1386 | |||
| AK130520.1 | - | 5 | 524 | |||
| BC101992.1 | - | 4 | 585 | 40023129 | ||

RNF150
Genomic Map