siRNA Details
| Gene Symbol: KIAA1522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198972.1 | NP_001185901.1 | 7 | 5121 | ||
| NM_001198973.1 | NP_001185902.1 | 5 | 2445 | |||
| NM_020888.2 | NP_065939.2 | 7 | 5206 | |||
| XM_006710791.3 | XP_006710854.1 | 7 | 5140 | |||
| XM_017001916.1 | XP_016857405.1 | 5 | 4804 | |||
| GenBank mRNA | AB040955.1 | - | 7 | 5088 | ||
| AL713671.1 | - | 2 | 2861 | |||
| AL833933.1 | - | 1 | 1511 | |||
| BC020306.1 | - | 1 | 1037 | |||

KIAA1522
Genomic Map