siRNA Details
| Gene Symbol: ZNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001203250.1 | NP_001190179.1 | 4 | 2326 | ||
| NM_021143.3 | NP_066966.2 | 4 | 2335 | |||
| GenBank mRNA | AK023094.1 | - | 4 | 2228 | ||
| Gene Symbol: ZNF625-ZNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037802.1 | 8 | 2692 | |||

ZNF20
Genomic Map