siRNA Details
Gene Symbol: Pappa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021362.1 | NP_067337.1 | 22 | 5636 | ||
GenBank mRNA | AF258461.1 | - | 21 | 4497 | ||
AF260433.1 | - | 10 | 1801 | |||
AF439513.1 | - | 22 | 5031 | |||
AF439514.1 | - | 23 | 5118 |
