siRNA Details
Gene Symbol: Fos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022197.2 | NP_071533.1 | 4 | 1039 | ||
GenBank mRNA | FQ225996.1 | - | 3 | 692 | ||
FQ228884.1 | - | 3 | 693 | |||
X06769.1 | - | 1021 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022197.2 | NP_071533.1 | 4 | 1039 | ||
GenBank mRNA | FQ225996.1 | - | 3 | 692 | ||
FQ228884.1 | - | 3 | 693 | |||
X06769.1 | - | 1021 |