siRNA Details
| Gene Symbol: Chchd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024166.6 | NP_077128.2 | 4 | 744 | ||
| GenBank mRNA | AK003399.1 | - | 4 | 579 | ||
| AK012691.1 | - | 3 | 570 | |||
| AK088374.1 | - | 4 | 745 | |||
| AK145492.1 | - | 4 | 562 | |||
| AK160762.1 | - | 4 | 563 | |||
| AK160779.1 | - | 4 | 564 | |||
| AK162133.1 | - | 4 | 562 | |||
| AK166796.1 | - | 4 | 564 | |||
| AK167593.1 | - | 4 | 564 | |||
| AK210686.1 | - | 2 | 92 | |||
| AK211922.1 | - | 1 | 93 | |||
| AK216042.1 | - | 2 | 92 | |||
| BC037682.1 | - | 3 | 1924 | |||
| BC042717.1 | - | 4 | 551 | 3154544 | ||
| BC069887.1 | - | 4 | 326 | |||
| BC082287.1 | - | 4 | 554 | 6397957 | ||
| BC083126.1 | - | 3 | 1887 | |||
| BC084682.1 | - | 4 | 533 | 6530688 | ||
| Gene Symbol: Gm6222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC158129.1 | - | 1 | 584 | 9007397 | |

Chchd2
Genomic Map