siRNA Details
| Gene Symbol: Stt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024222.2 | NP_077184.2 | 15 | 2717 | ||
| GenBank mRNA | AK012153.1 | - | 15 | 2387 | ||
| AK018758.2 | - | 15 | 2427 | |||
| AK145674.1 | - | 15 | 2358 | |||
| AK148065.1 | - | 15 | 2416 | |||
| AK152899.1 | - | 15 | 2765 | |||
| AK154979.1 | - | 15 | 2406 | |||
| AK170079.1 | - | 15 | 2429 | |||
| AX799084.1 | - | 15 | 2428 | |||
| BC003206.1 | - | 10 | 1429 | |||
| BC013054.1 | - | 5 | 835 | |||
| BC052433.1 | - | 15 | 2717 | 6837097 | ||

Stt3b
Genomic Map