siRNA Details
Gene Symbol: Ypel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025347.2 | NP_079623.1 | 5 | 906 | ||
NM_026875.2 | NP_081151.2 | 5 | 1006 | |||
GenBank mRNA | AA797224.1 | - | 1 | 366 | ||
AF523353.1 | - | 5 | 896 | |||
AK002925.1 | - | 5 | 890 | |||
AK003371.1 | - | 5 | 907 | |||
AK004431.1 | - | 5 | 979 | |||
BC009171.1 | - | 5 | 880 |
