siRNA Details
| Gene Symbol: Pdcd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026549.3 | NP_080825.1 | 5, 6 | 827 | ||
| GenBank mRNA | AK003339.1 | - | 5, 6 | 836 | ||
| AK077948.1 | - | 5, 6 | 828 | |||
| AK151495.1 | - | 5, 6 | 831 | |||
| AK154811.1 | - | 5, 6 | 843 | |||
| BC025090.1 | - | 2, 3 | 291 | |||
| BC038524.1 | - | 2, 3 | 297 | |||
| BC115797.1 | - | 5, 6 | 826 | 40103840 | ||

Pdcd2l
Genomic Map