siRNA Details
| Gene Symbol: Tmem126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026734.1 | NP_081010.1 | 2, 3 | 217 | ||
| GenBank mRNA | AK003201.1 | - | 2, 3 | 218 | ||
| BC049680.1 | - | 2, 3 | 217 | 6596401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem126b| Gene Symbol: Tmem126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026734.1 | NP_081010.1 | 2, 3 | 217 | ||
| GenBank mRNA | AK003201.1 | - | 2, 3 | 218 | ||
| BC049680.1 | - | 2, 3 | 217 | 6596401 | ||