siRNA Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199187.2 | NP_001186116.1 | 6 | 623 | ||
| NM_026937.3 | NP_081213.1 | 6 | 636 | |||
| GenBank mRNA | AK007519.1 | - | 6 | 620 | ||
| AK139944.1 | - | 6 | 623 | |||
| AK179536.1 | - | 2 | 136 | |||
| BC030905.1 | - | 6 | 602 | 4503445 | ||

Ascc1
Genomic Map