siRNA Details
| Gene Symbol: Draxin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027426.3 | NP_081702.2 | 7 | 1739 | ||
| GenBank mRNA | AK011831.1 | - | 4 | 904 | ||
| AK086198.1 | - | 7 | 1740 | |||
| BC058734.1 | - | 3 | 855 | |||
| BC060239.1 | - | 7 | 1720 | 6853328 | ||

Draxin
Genomic Map