siRNA Details
Gene Symbol: Pnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027438.3 | NP_081714.2 | 1 | 2128 | ||
GenBank mRNA | AK017476.1 | - | 1 | 1922 | ||
AK028331.1 | - | 1 | 2066 | |||
AK136933.1 | - | 1 | 2130 | |||
BM939442.1 | - | 1 | 81 |
