siRNA Details
| Gene Symbol: Fam81a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029784.2 | NP_084060.1 | 8 | 1120 | ||
| GenBank mRNA | AK018282.1 | - | 8 | 1077 | ||
| AK038532.1 | - | 8 | 1079 | |||
| AK044090.1 | - | 8 | 1120 | |||
| AK046408.1 | - | 8 | 1013 | |||
| AK079541.1 | - | 8 | 1080 | |||
| AK135091.1 | - | 7 | 835 | |||
| BC025646.1 | - | 8 | 1146 | 5324986 | ||

Fam81a
Genomic Map