siRNA Details
| Gene Symbol: Fam81a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029784.2 | NP_084060.1 | 9 | 2776 | ||
| GenBank mRNA | AK044090.1 | - | 9 | 2771 | ||
| AK149206.1 | - | 657 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam81a| Gene Symbol: Fam81a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029784.2 | NP_084060.1 | 9 | 2776 | ||
| GenBank mRNA | AK044090.1 | - | 9 | 2771 | ||
| AK149206.1 | - | 657 | ||||