siRNA Details
| Gene Symbol: Rnf170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029965.2 | NP_084241.1 | 8 | 1445 | ||
| GenBank mRNA | AK036051.1 | - | 8 | 1305 | ||
| AK162554.1 | - | 8 | 1463 | |||
| BC031383.1 | - | 8 | 1229 | |||
| BC114210.1 | - | 8 | 1445 | 6309632 | ||
| BC138930.1 | - | 8 | 1272 | 8861952 | ||
| BC145718.1 | - | 8 | 1277 | 40131021 | ||

Rnf170
Genomic Map