siRNA Details
| Gene Symbol: Ulbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029975.2 | NP_084251.1 | 3 | 657 | ||
| GenBank mRNA | AK020784.1 | - | 3 | 532 | ||
| AK037408.1 | - | 3 | 629 | |||
| AK084712.1 | - | 756 | ||||
| AK085119.1 | - | 819 | ||||
| AY249021.1 | - | 2 | 286 | |||
| BC132070.1 | - | 3 | 657 | 40130347 | ||
| BC132072.1 | - | 3 | 657 | 40130349 | ||
| EU047812.1 | - | 3 | 521 | |||

Ulbp1
Genomic Map