siRNA Details
| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020026.1 | - | 4 | 498 | ||
| AK046610.1 | - | 4 | 526 | |||
| AK089083.1 | - | 4 | 339 | |||
| AK168294.1 | - | 5 | 786 | |||
| BC031464.1 | - | 4 | 483 | 4017482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc6b| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020026.1 | - | 4 | 498 | ||
| AK046610.1 | - | 4 | 526 | |||
| AK089083.1 | - | 4 | 339 | |||
| AK168294.1 | - | 5 | 786 | |||
| BC031464.1 | - | 4 | 483 | 4017482 | ||