siRNA Details
| Gene Symbol: 1700034E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030097.1 | NP_084373.1 | 4 | 378 | ||
| GenBank mRNA | AK018840.1 | - | 3 | 338 | ||
| BC048563.1 | - | 4 | 378 | 6704020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700034E13Rik| Gene Symbol: 1700034E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030097.1 | NP_084373.1 | 4 | 378 | ||
| GenBank mRNA | AK018840.1 | - | 3 | 338 | ||
| BC048563.1 | - | 4 | 378 | 6704020 | ||