siRNA Details
| Gene Symbol: 8030411F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030135.2 | NP_084411.1 | 3 | 450 | ||
| GenBank mRNA | AF440735.1 | - | 3 | 450 | ||
| AK020193.1 | - | 3 | 366 | |||
| BC132218.1 | - | 3 | 395 | 40130495 | ||
| BC132222.1 | - | 3 | 395 | 40130499 | ||
| BC145532.1 | - | 3 | 359 | |||

8030411F24Rik
Genomic Map