siRNA Details
| Gene Symbol: Itga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030842.1 | NP_110469.1 | 13, 14 | 1931 | ||
| XM_006240800.2 | XP_006240862.1 | 13, 14 | 2121 | |||
| XM_008765044.1 | XP_008763266.1 | 10, 11 | 1571 | |||
| GenBank mRNA | BC088846.1 | - | 13, 14 | 1931 | 7313230 | |
| X65036.1 | - | 13, 14 | 1804 | |||

Itga7
Genomic Map