siRNA Details
| Gene Symbol: Acvr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031554.1 | NP_113742.1 | 10 | 1532 | ||
| XM_006244072.2 | XP_006244134.1 | 9 | 1634 | |||
| GenBank mRNA | BC097358.1 | - | 10 | 1532 | 7442221 | |
| L10640.1 | - | 8 | 976 | |||
| M87067.1 | - | 10 | 1774 | |||

Acvr2b
Genomic Map