siRNA Details
| Gene Symbol: Elmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080288.2 | NP_525027.1 | 18, 19 | 2055 | ||
| NM_198093.3 | NP_932761.1 | 3, 4 | 578 | |||
| GenBank mRNA | AF398883.1 | - | 17, 18 | 1719 | ||
| AK032085.1 | - | 3, 4 | 488 | |||
| AK053306.1 | - | 3, 4 | 489 | |||
| AK220253.1 | - | 18, 19 | 1943 | |||
| BC024727.1 | - | 3, 4 | 475 | |||
| BC031782.1 | - | 7, 8 | 793 | 3990906 | ||

Elmo1
Genomic Map