siRNA Details
| Gene Symbol: Elmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080288.2 | NP_525027.1 | 17 | 1927 | ||
| NM_198093.3 | NP_932761.1 | 2 | 450 | |||
| GenBank mRNA | AF398883.1 | - | 16 | 1591 | ||
| AK032085.1 | - | 2 | 360 | |||
| AK053306.1 | - | 2 | 361 | |||
| AK220253.1 | - | 17 | 1815 | |||
| BC024727.1 | - | 2 | 347 | |||
| BC031782.1 | - | 6 | 665 | 3990906 | ||

Elmo1
Genomic Map