siRNA Details
| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080402.1 | NP_536327.1 | 6 | 713 | ||
| XM_008766473.1 | XP_008764695.1 | 6 | 713 | |||
| GenBank mRNA | AY669340.1 | - | 6 | 713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ryk| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080402.1 | NP_536327.1 | 6 | 713 | ||
| XM_008766473.1 | XP_008764695.1 | 6 | 713 | |||
| GenBank mRNA | AY669340.1 | - | 6 | 713 | ||