siRNA Details
| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134367.1 | NP_599194.1 | 2 | 261 | ||
| XM_008770813.1 | XP_008769035.1 | 2 | 444 | |||
| XM_008770814.1 | XP_008769036.1 | 2 | 443 | |||
| XM_008770815.1 | XP_008769037.1 | 2 | 878 | |||
| XM_008770816.1 | XP_008769038.1 | 3 | 255 | |||
| XM_008770817.1 | XP_008769039.1 | 2 | 792 | |||
| GenBank mRNA | BC079027.1 | - | 2 | 261 | 7114037 | |
| GQ376198.1 | - | 2 | 170 | |||
| GQ376199.1 | - | 2 | 170 | |||
| GQ376200.1 | - | 2 | 170 | |||

Ppp3cc
Genomic Map