siRNA Details
| Gene Symbol: CCDC167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138493.2 | NP_612502.1 | 4 | 475 | ||
| GenBank mRNA | AF161383.1 | - | 4 | 474 | ||
| BC003515.1 | - | 4 | 420 | |||
| BC108655.1 | - | 4 | 448 | 6053765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC167| Gene Symbol: CCDC167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138493.2 | NP_612502.1 | 4 | 475 | ||
| GenBank mRNA | AF161383.1 | - | 4 | 474 | ||
| BC003515.1 | - | 4 | 420 | |||
| BC108655.1 | - | 4 | 448 | 6053765 | ||